RIE156319115 Rieke Metals, Inc.

2-[(4-Methylpiperazino)methyl]phenylmagnesium bromide

  • MDL number MFCD07698758
  • Molecular Weight : 293.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3190

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3190
Availability details for
Quantity
  • Availability
  • Add to Cart