RIE156319117 Rieke Metals, Inc.

4-[(4-Methylpiperazino)methyl]phenylmagnesium bromide

  • MDL number MFCD07698760
  • Molecular Weight : 293.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3192

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3192
Availability details for
Quantity
  • Availability
  • Add to Cart