RIE156319118 Rieke Metals, Inc.

4-(1,2,3,4-Tetrahydroquinolino-2-ylmethyl)phenylmagnesium bromide

  • MDL number MFCD07698761
  • Molecular Weight : 326.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3193

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3193
Availability details for
Quantity
  • Availability
  • Add to Cart