RIE156319121 Rieke Metals, Inc.

3-Chloro-4-methylphenylmagnesium iodide

  • MDL number MFCD07698764
  • Molecular Weight : 276.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3196

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3196
Availability details for
Quantity
  • Availability
  • Add to Cart