RIE156319122 Rieke Metals, Inc.

4-Chlorophenethylmagnesium bromide

  • MDL number MFCD07698765
  • Cas Number : 36159-18-9
  • Molecular Weight : 243.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3197

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3197
Availability details for
Quantity
  • Availability
  • Add to Cart