RIE156319126 Rieke Metals, Inc.

4-Methylphenethylmagnesium bromide

  • MDL number MFCD07698769
  • Molecular Weight : 223.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart