RIE156319128 Rieke Metals, Inc.

2-Methylphenethylmagnesium bromide

  • MDL number MFCD07698771
  • Molecular Weight : 223.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3203
Availability details for
Quantity
  • Availability
  • Add to Cart