RIE156319129 Rieke Metals, Inc.

4-Methoxyphenethylmagnesium bromide

  • MDL number MFCD07698772
  • Cas Number : 36278-54-3
  • Molecular Weight : 239.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3204

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3204
Availability details for
Quantity
  • Availability
  • Add to Cart