RIE156319130 Rieke Metals, Inc.

3-Methoxyphenethylmagnesium bromide

  • MDL number MFCD07698773
  • Cas Number : 125159-92-4
  • Molecular Weight : 239.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3205

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3205
Availability details for
Quantity
  • Availability
  • Add to Cart