RIE156319131 Rieke Metals, Inc.

2-Methoxyphenethylmagnesium bromide

  • MDL number MFCD07698774
  • Cas Number : 123427-77-0
  • Molecular Weight : 239.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3206

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3206
Availability details for
Quantity
  • Availability
  • Add to Cart