RIE156319133 Rieke Metals, Inc.

3-Chloro-5-fluorobenzylmagnesium chloride

  • MDL number MFCD07698775
  • Molecular Weight : 203.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3209

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3209
Availability details for
Quantity
  • Availability
  • Add to Cart