RIE156319135 Rieke Metals, Inc.

3,5-Dimethoxyphenylmagnesium bromide

  • MDL number MFCD08064086
  • Cas Number : 322640-05-1
  • Molecular Weight : 241.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3212

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3212
Availability details for
Quantity
  • Availability
  • Add to Cart