RIE156319149 Rieke Metals, Inc.

3,4'-dibromobenzophenone

  • MDL number MFCD00672016
  • Cas Number : 83699-51-8
  • Molecular Weight : 340.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5004

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5004
Availability details for
Quantity
  • Availability
  • Add to Cart