RIE156319150 Rieke Metals, Inc.

2,4'-Dibromobenzophenone

  • MDL number MFCD00672017
  • Cas Number : 78281-59-1
  • Molecular Weight : 340.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5005

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5005
Availability details for
Quantity
  • Availability
  • Add to Cart