RIE156319152 Rieke Metals, Inc.

2-Bromo-4'-cyanobenzophenone

  • MDL number MFCD00672019
  • Cas Number : 746651-77-4
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5007

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5007
Availability details for
Quantity
  • Availability
  • Add to Cart