RIE156319153 Rieke Metals, Inc.

4-bromo-4'-cyanobenzophenone

  • MDL number MFCD00672021
  • Cas Number : 452929-54-3
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5009

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5009
Availability details for
Quantity
  • Availability
  • Add to Cart