RIE156319155 Rieke Metals, Inc.

3-Bromo-4'carboethoxybenzophenone

  • MDL number MFCD00672023
  • Cas Number : 746651-79-6
  • Molecular Weight : 333.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5012

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5012
Availability details for
Quantity
  • Availability
  • Add to Cart