RIE156319156 Rieke Metals, Inc.

4-Bromo-4'carboethoxybenzophenone

  • MDL number MFCD00672024
  • Cas Number : 746651-80-9
  • Molecular Weight : 333.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5013

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5013
Availability details for
Quantity
  • Availability
  • Add to Cart