RIE156319159 Rieke Metals, Inc.

2-Bromo-2'-cyanobenzophenone

  • MDL number MFCD00672029
  • Cas Number : 746651-83-2
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-bromobenzoyl)benzonitrile
Inchi Code 1S/C14H8BrNO/c15-13-8-4-3-7-12(13)14(17)11-6-2-1-5-10(11)9-16/h1-8H
Inchi Key RAMDLDMVUHKNBK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5019

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5019
Availability details for
Quantity
  • Availability
  • Add to Cart