RIE156319160 Rieke Metals, Inc.

3-Bromo-2'-cyanobenzophenone

  • MDL number MFCD00672030
  • Cas Number : 746651-84-3
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5020

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5020
Availability details for
Quantity
  • Availability
  • Add to Cart