RIE156319162 Rieke Metals, Inc.

2-bromo-5-methoxybenzophenone

  • MDL number MFCD00672036
  • Cas Number : 60080-98-0
  • Molecular Weight : 291.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5026

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5026
Availability details for
Quantity
  • Availability
  • Add to Cart