RIE156319163 Rieke Metals, Inc.

2,4'-Dibromo-5-methoxybenzophenone

  • MDL number MFCD00672037
  • Cas Number : 890098-03-0
  • Molecular Weight : 370.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5027

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5027
Availability details for
Quantity
  • Availability
  • Add to Cart