RIE156319164 Rieke Metals, Inc.

2,3'-dibromo-5-methoxybenzophenone

  • MDL number MFCD00672038
  • Cas Number : 746651-87-6
  • Molecular Weight : 370.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5028

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5028
Availability details for
Quantity
  • Availability
  • Add to Cart