RIE156319169 Rieke Metals, Inc.

2-chloro-3',4',5',-trifluorobenzophenone

  • MDL number MFCD01319930
  • Cas Number : 746651-94-5
  • Molecular Weight : 270.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5043

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5043
Availability details for
Quantity
  • Availability
  • Add to Cart