RIE156319170 Rieke Metals, Inc.

2-chloro-3',5'-difluorobenzophenone

  • MDL number MFCD01319933
  • Cas Number : 746651-97-8
  • Molecular Weight : 252.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5046

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5046
Availability details for
Quantity
  • Availability
  • Add to Cart