RIE156319171 Rieke Metals, Inc.

2-Methoxy-3'-methylbenzophenone

  • MDL number MFCD01311525
  • Cas Number : 33785-70-5
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-03
Availability details for
Quantity
  • Availability
  • Add to Cart