RIE156319172 Rieke Metals, Inc.

3,2'-Dimethoxybenzophenone

  • MDL number MFCD01311507
  • Cas Number : 21554-74-5
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-06
Availability details for
Quantity
  • Availability
  • Add to Cart