RIE156319174 Rieke Metals, Inc.

3-Cyano-2'-methoxybenzophenone

  • MDL number MFCD01311537
  • Cas Number : 131117-96-9
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-09
Availability details for
Quantity
  • Availability
  • Add to Cart