RIE156319179 Rieke Metals, Inc.

2-Methoxy-2'-thiomethylbenzophenone

  • MDL number MFCD07698783
  • Cas Number : 746652-03-9
  • Molecular Weight : 258.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-14
Availability details for
Quantity
  • Availability
  • Add to Cart