RIE156319181 Rieke Metals, Inc.

2-Ethyl-2'-methoxybenzophenone

  • MDL number MFCD01311518
  • Cas Number : 750633-44-4
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-ethylphenyl)(2-methoxyphenyl)methanone
Inchi Code 1S/C16H16O2/c1-3-12-8-4-5-9-13(12)16(17)14-10-6-7-11-15(14)18-2/h4-11H,3H2,1-2H3
Inchi Key JSOMBRTXVWAKFH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5049-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-16
Availability details for
Quantity
  • Availability
  • Add to Cart