RIE156319182 Rieke Metals, Inc.

4-Ethyl-2'-methoxybenzophenone

  • MDL number MFCD01311519
  • Cas Number : 82520-38-5
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-17
Availability details for
Quantity
  • Availability
  • Add to Cart