RIE156319201 Rieke Metals, Inc.

2-fluoro-2'-methoxybenzophenone

  • MDL number MFCD07698787
  • Cas Number : 890098-05-2
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluorophenyl)(2-methoxyphenyl)methanone
Inchi Code 1S/C14H11FO2/c1-17-13-9-5-3-7-11(13)14(16)10-6-2-4-8-12(10)15/h2-9H,1H3
Inchi Key CQZHJHDLSUCHIU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5049-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-37
Availability details for
Quantity
  • Availability
  • Add to Cart