RIE156319203 Rieke Metals, Inc.

2-Cyano-3'-methoxybenzophenone

  • MDL number MFCD01311568
  • Cas Number : 750633-58-0
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-methoxybenzoyl)benzonitrile
Inchi Code 1S/C15H11NO2/c1-18-13-7-4-6-11(9-13)15(17)14-8-3-2-5-12(14)10-16/h2-9H,1H3
Inchi Key QOYOOSYWDUDBLL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-08
Availability details for
Quantity
  • Availability
  • Add to Cart