RIE156319208 Rieke Metals, Inc.

3-Methoxy-2'-thiomethylbenzophenone

  • MDL number MFCD07698788
  • Cas Number : 750633-63-7
  • Molecular Weight : 258.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-14
Availability details for
Quantity
  • Availability
  • Add to Cart