RIE156319210 Rieke Metals, Inc.

2-ethyl-3'-methoxybenzophenone

  • MDL number MFCD01311550
  • Cas Number : 750633-65-9
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-16
Availability details for
Quantity
  • Availability
  • Add to Cart