RIE156319211 Rieke Metals, Inc.

3-Fluoro-3'-methoxybenzophenone

  • MDL number MFCD01311552
  • Cas Number : 120553-05-1
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-22
Availability details for
Quantity
  • Availability
  • Add to Cart