RIE156319215 Rieke Metals, Inc.

2,6-dimethyl-3'-methoxybenzophenone

  • MDL number MFCD01311544
  • Cas Number : 750633-72-8
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-29
Availability details for
Quantity
  • Availability
  • Add to Cart