RIE156319217 Rieke Metals, Inc.

4-bromo-3-fluoro-3'-methoxybenzophenone

  • MDL number MFCD01311554
  • Cas Number : 750633-75-1
  • Molecular Weight : 309.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-32
Availability details for
Quantity
  • Availability
  • Add to Cart