RIE156319218 Rieke Metals, Inc.

4-chloro-3-fluoro-3'-methoxybenzophenone

  • MDL number MFCD01311565
  • Cas Number : 750633-76-2
  • Molecular Weight : 264.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-33
Availability details for
Quantity
  • Availability
  • Add to Cart