RIE156319219 Rieke Metals, Inc.

3-chloro-4-fluoro-3'-methoxybenzophenone

  • MDL number MFCD01311564
  • Cas Number : 750633-77-3
  • Molecular Weight : 264.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-34
Availability details for
Quantity
  • Availability
  • Add to Cart