RIE156319230 Rieke Metals, Inc.

4-methoxy-4'-n-pentylbenzophenone

  • MDL number MFCD01311585
  • Cas Number : 64357-56-8
  • Molecular Weight : 282.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-25
Availability details for
Quantity
  • Availability
  • Add to Cart