RIE156319231 Rieke Metals, Inc.

2,3-dimethyl-4'-methoxybenzophenone

  • MDL number MFCD01311571
  • Cas Number : 134994-51-7
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-26
Availability details for
Quantity
  • Availability
  • Add to Cart