RIE156319233 Rieke Metals, Inc.

3,5-Dimethyl-4'-methoxybenzophenone

  • MDL number MFCD01311575
  • Cas Number : 10472-83-0
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-31
Availability details for
Quantity
  • Availability
  • Add to Cart