RIE156319235 Rieke Metals, Inc.

4-chloro-3-fluoro-4'-methoxybenzophenone

  • MDL number MFCD01311590
  • Cas Number : 760192-86-7
  • Molecular Weight : 264.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-33
Availability details for
Quantity
  • Availability
  • Add to Cart