RIE156319237 Rieke Metals, Inc.

4-bromophenyl-2'-naphthyl ketone

  • MDL number MFCD01319940
  • Cas Number : 760192-88-9
  • Molecular Weight : 311.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(2-naphthyl)methanone
Inchi Code 1S/C17H11BrO/c18-16-9-7-13(8-10-16)17(19)15-6-5-12-3-1-2-4-14(12)11-15/h1-11H
Inchi Key RPYONPWJLGOFGD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5057

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5057
Availability details for
Quantity
  • Availability
  • Add to Cart