RIE156319239 Rieke Metals, Inc.

2-Chlorobenzophenone ethylene ketal

  • MDL number MFCD02260356
  • Cas Number : 760192-90-3
  • Molecular Weight : 260.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5062

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5062
Availability details for
Quantity
  • Availability
  • Add to Cart