RIE156319241 Rieke Metals, Inc.

3'-bromo-2-iodobenzophenone

  • MDL number MFCD02260360
  • Cas Number : 890098-10-9
  • Molecular Weight : 387.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5069

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5069
Availability details for
Quantity
  • Availability
  • Add to Cart