RIE156319242 Rieke Metals, Inc.

4'-bromo-2-iodobenzophenone

  • MDL number MFCD02260361
  • Cas Number : 890098-11-0
  • Molecular Weight : 387.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(2-iodophenyl)methanone
Inchi Code 1S/C13H8BrIO/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8H
Inchi Key QMGDAKSOOZSSLX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5070

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5070
Availability details for
Quantity
  • Availability
  • Add to Cart