RIE156319245 Rieke Metals, Inc.

3-Bromo-4'-iodobenzophenone

  • MDL number MFCD02260364
  • Cas Number : 890098-13-2
  • Molecular Weight : 387.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(4-iodophenyl)methanone
Inchi Code 1S/C13H8BrIO/c14-11-3-1-2-10(8-11)13(16)9-4-6-12(15)7-5-9/h1-8H
Inchi Key UZSFJVSPCZOCKL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5073

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5073
Availability details for
Quantity
  • Availability
  • Add to Cart