RIE156319247 Rieke Metals, Inc.

4'-Chloro-2-iodobenzophenone

  • MDL number MFCD02260367
  • Cas Number : 99847-66-2
  • Molecular Weight : 342.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5077

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5077
Availability details for
Quantity
  • Availability
  • Add to Cart